siRNA Details
Gene Symbol: Olfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038612.1 | NP_001033701.1 | 2, 3 | 570 | ||
NM_001038613.1 | NP_001033702.1 | 2, 3 | 529 | |||
NM_001038614.1 | NP_001033703.1 | 2, 3 | 529 | |||
NM_019498.2 | NP_062371.2 | 2, 3 | 570 | |||
GenBank mRNA | AF028740.1 | - | 2, 3 | 225 | ||
AK003031.1 | - | 2, 3 | 462 | |||
AK147618.1 | - | 1 | 5419 | |||
BC026547.1 | - | 2, 3 | 371 | 5036675 | ||
BQ768981.1 | - | 2, 3 | 529 | |||
BU512660.1 | - | 2, 3 | 575 | |||
CB203212.1 | - | 2, 3 | 205 | |||
CT010369.1 | - | 2, 3 | 309 | |||
D78262.1 | - | 2, 3 | 583 | |||
D78263.1 | - | 2, 3 | 583 | |||
D78264.1 | - | 2, 3 | 630 | |||
D78265.1 | - | 2, 3 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
