siRNA Details
Gene Symbol: Rhog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019566.3 | NP_062512.1 | 2 | 847 | ||
GenBank mRNA | AK143623.1 | - | 2 | 840 | ||
AK149848.1 | - | 2 | 840 | |||
AK169853.1 | - | 2 | 831 | |||
AK188210.1 | - | 1 | 325 | |||
BC059775.1 | - | 2 | 826 | 5370916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
