siRNA Details
Gene Symbol: Stx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024414.2 | NP_077725.1 | 4, 5 | 522 | ||
GenBank mRNA | BC132042.1 | - | 4, 5 | 317 | 40130319 | |
BC132068.1 | - | 4, 5 | 317 | 40130345 | ||
D29743.1 | - | 4, 5 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
