siRNA Details
Gene Symbol: KRT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117465.1 | - | 6 | 771 | 40126016 | |
BC117467.1 | - | 6 | 772 | 40126018 | ||
X81420.1 | - | 6 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
