siRNA Details
Gene Symbol: Hdac7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204275.1 | NP_001191204.1 | 14 | 2253 | ||
NM_001204276.1 | NP_001191205.1 | 15 | 2301 | |||
NM_001204277.1 | NP_001191206.1 | 14 | 2172 | |||
NM_001204278.1 | NP_001191207.1 | 13 | 2142 | |||
NM_001204279.1 | NP_001191208.1 | 13 | 2070 | |||
NM_001204280.1 | NP_001191209.1 | 13 | 2142 | |||
NM_001204281.1 | NP_001191210.1 | 12 | 1705 | |||
NM_019572.3 | NP_062518.2 | 13 | 2154 | |||
GenBank mRNA | AF207749.1 | - | 13 | 1764 | ||
AK028515.1 | - | 12 | 1678 | |||
AK030863.1 | - | 14 | 2054 | |||
AK036586.1 | - | 15 | 2029 | |||
AK037282.1 | - | 13 | 1806 | |||
AK040406.1 | - | 13 | 1856 | |||
AK044287.1 | - | 14 | 1900 | |||
AK088828.1 | - | 13 | 1838 | |||
AK088945.1 | - | 13 | 1898 | |||
AK131123.1 | - | 13 | 1881 | |||
AK146315.1 | - | 13 | 1943 | |||
AK150041.1 | - | 13 | 1865 | |||
BC057332.1 | - | 13 | 2058 | 6826982 |

More Information
Other Species Targeted

Gene Ontology Categories:
