siRNA Details
Gene Symbol: Hdac7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204275.1 | NP_001191204.1 | 5 | 894 | ||
NM_001204276.1 | NP_001191205.1 | 6 | 963 | |||
NM_001204277.1 | NP_001191206.1 | 5 | 834 | |||
NM_001204278.1 | NP_001191207.1 | 5 | 894 | |||
NM_001204279.1 | NP_001191208.1 | 5 | 822 | |||
NM_001204280.1 | NP_001191209.1 | 5 | 894 | |||
NM_001204281.1 | NP_001191210.1 | 5 | 652 | |||
NM_019572.3 | NP_062518.2 | 5 | 906 | |||
GenBank mRNA | AF207749.1 | - | 5 | 516 | ||
AK028515.1 | - | 5 | 625 | |||
AK036586.1 | - | 6 | 691 | |||
AK037282.1 | - | 5 | 557 | |||
AK040406.1 | - | 5 | 608 | |||
AK044287.1 | - | 5 | 562 | |||
AK088828.1 | - | 5 | 590 | |||
AK088945.1 | - | 5 | 650 | |||
AK131123.1 | - | 5 | 633 | |||
AK146315.1 | - | 5 | 694 | |||
AK150041.1 | - | 5 | 617 | |||
AK206054.1 | - | 1 | 14 | |||
AK210276.1 | - | 1 | 14 | |||
AK213805.1 | - | 1 | 14 | |||
AK216333.1 | - | 1 | 14 | |||
AK219912.1 | - | 1 | 14 | |||
BC057332.1 | - | 5 | 810 | 6826982 |
