siRNA Details
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019635.2 | NP_062609.2 | 3 | 430 | ||
GenBank mRNA | AF271361.1 | - | 3 | 176 | ||
AY058922.1 | - | 3 | 316 | |||
BC049123.1 | - | 3 | 304 | 6813956 | ||
U28726.1 | - | 3 | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
