siRNA Details
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019635.2 | NP_062609.2 | 4 | 587 | ||
GenBank mRNA | AF271361.1 | - | 4 | 333 | ||
AY058922.1 | - | 4 | 473 | |||
BC037440.1 | - | 2 | 343 | 3585312 | ||
BC049123.1 | - | 4 | 461 | 6813956 | ||
U28726.1 | - | 4 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
