siRNA Details
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019814.4 | NP_062788.1 | 3 | 635 | ||
GenBank mRNA | AB253941.1 | - | 2 | 147 | ||
AF141312.1 | - | 3 | 166 | |||
AK004166.1 | - | 3 | 262 | |||
AK008756.1 | - | 3 | 262 | |||
AK018360.1 | - | 3 | 263 | |||
AY028386.1 | - | 3 | 188 | |||
AY028387.1 | - | 3 | 173 | |||
BC021594.1 | - | 3 | 185 | 5148784 | ||
BC090259.1 | - | 3 | 239 | 5060413 | ||
BC132371.1 | - | 3 | 253 | 40130648 | ||
BC151191.1 | - | 3 | 253 | 8861324 | ||
BE457620.1 | - | 3 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
