siRNA Details
Gene Symbol: Mycbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019660.3 | NP_062634.2 | 5 | 1187 | ||
GenBank mRNA | AB015858.1 | - | 5 | 1146 | ||
AK037661.1 | - | 1197 | ||||
AK077633.1 | - | 5 | 1206 | |||
AK077762.1 | - | 5 | 1191 | |||
AK077995.1 | - | 5 | 1191 | |||
AK132198.1 | - | 1052 | ||||
AK145773.1 | - | 5 | 1188 | |||
BC028344.1 | - | 5 | 1184 | 3985166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
