siRNA Details
Gene Symbol: Mycbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019660.3 | NP_062634.2 | 4 | 285 | ||
GenBank mRNA | AB015858.1 | - | 4 | 248 | ||
AK033289.1 | - | 4 | 385 | |||
AK037661.1 | - | 295 | ||||
AK077633.1 | - | 4 | 304 | |||
AK077762.1 | - | 4 | 289 | |||
AK077995.1 | - | 4 | 289 | |||
AK132198.1 | - | 150 | ||||
AK145773.1 | - | 4 | 286 | |||
BC028344.1 | - | 4 | 283 | 3985166 | ||
BC048478.1 | - | 4 | 466 | 6774657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
