siRNA Details
Gene Symbol: Pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019746.4 | NP_062720.1 | 4 | 351 | ||
GenBank mRNA | AF161074.1 | - | 4 | 240 | ||
AK007297.1 | - | 4 | 264 | |||
AK008632.1 | - | 4 | 365 | |||
BC048476.1 | - | 4 | 251 | 6773378 | ||
BC056167.1 | - | 4 | 243 | 5712632 | ||
BC092092.1 | - | 4 | 351 | 3492310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
