siRNA Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019795.4 | NP_062769.2 | 4 | 937 | ||
GenBank mRNA | AB028861.1 | - | 4 | 385 | ||
AK011875.1 | - | 4 | 376 | |||
AK076032.1 | - | 4 | 408 | |||
AK145506.1 | - | 4 | 399 | |||
AK145739.1 | - | 4 | 399 | |||
AK166872.1 | - | 4 | 399 | |||
AY323828.1 | - | 4 | 373 | |||
BC023681.1 | - | 4 | 581 | 5343384 | ||
BC055729.1 | - | 4 | 936 | 6414499 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_213625, XM_006247313, XM_006247312, XM_006247311, FQ212948, and others.
Gene Ontology Categories:
