siRNA Details
Gene Symbol: Sult1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019878.4 | NP_063931.1 | 4 | 443 | ||
GenBank mRNA | AF022894.1 | - | 4 | 462 | ||
AK033498.1 | - | 4 | 446 | |||
AK087595.1 | - | 4 | 479 | |||
BC024361.1 | - | 3 | 402 | 4990345 | ||
U92076.1 | - | 4 | 388 |
