siRNA Details
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019790.4 | NP_062764.1 | 1, 2 | 595 | ||
GenBank mRNA | AB017270.1 | - | 1, 2 | 558 | ||
AB041565.1 | - | 1, 2 | 599 | |||
AK029307.1 | - | 1, 2 | 595 | |||
AK030053.1 | - | 6, 7 | 1831 | |||
AK076254.1 | - | 1, 2 | 595 | |||
AK135038.1 | - | 1, 2 | 582 | |||
BC034850.1 | - | 1, 2 | 571 | 1397175 |

More Information
Other Species Targeted

