siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 8 | 1825 | ||
NM_019831.3 | NP_062805.1 | 8 | 1685 | |||
GenBank mRNA | AF156605.1 | - | 8 | 2311 | ||
AK122268.1 | - | 7 | 1990 | |||
BC050131.1 | - | 8 | 1709 | |||
BC139001.1 | - | 7 | 1618 | 8862023 | ||
BC145497.1 | - | 7 | 1612 | 9054060 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001040155.
Gene Ontology Categories:
