siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 18, 19 | 3251 | ||
NM_019831.3 | NP_062805.1 | 18, 19 | 3111 | |||
GenBank mRNA | AF156605.1 | - | 18, 19 | 3737 | ||
AK122268.1 | - | 16, 17 | 3627 | |||
BC050131.1 | - | 18, 19 | 3134 | |||
BC139001.1 | - | 17, 18 | 3044 | 8862023 | ||
BC145497.1 | - | 17, 18 | 3038 | 9054060 |

More Information
Other Species Targeted

Gene Ontology Categories:
