siRNA Details
Gene Symbol: Zmym3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177985.1 | NP_001171456.1 | 12 | 2367 | ||
NM_019831.3 | NP_062805.1 | 12 | 2227 | |||
GenBank mRNA | AF156605.1 | - | 12 | 2853 | ||
AK122268.1 | - | 11 | 2532 | |||
BC050131.1 | - | 12 | 2250 | |||
BC139001.1 | - | 11 | 2160 | 8862023 | ||
BC145497.1 | - | 11 | 2154 | 9054060 |

More Information
Other Species Targeted

Gene Ontology Categories:
