siRNA Details
Gene Symbol: 1110004F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019772.2 | NP_062746.1 | 3 | 247 | ||
GenBank mRNA | AB025407.1 | - | 3 | 247 | ||
AK003415.1 | - | 3 | 288 | |||
AK144059.1 | - | 3 | 274 | |||
AK167571.1 | - | 3 | 276 | |||
AK168206.1 | - | 3 | 281 | |||
AK169309.1 | - | 3 | 274 | |||
AK180958.1 | - | 3 | 125 | |||
AK182617.1 | - | 3 | 125 | |||
BC072575.1 | - | 3 | 283 | 5715413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
