siRNA Details
Gene Symbol: 1110004F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019772.2 | NP_062746.1 | 2 | 182 | ||
GenBank mRNA | AB025407.1 | - | 2 | 182 | ||
AK003415.1 | - | 2 | 223 | |||
AK144059.1 | - | 2 | 209 | |||
AK167571.1 | - | 2 | 211 | |||
AK168206.1 | - | 2 | 216 | |||
AK169309.1 | - | 2 | 209 | |||
AK180958.1 | - | 2 | 60 | |||
BC072575.1 | - | 2 | 218 | 5715413 |
