siRNA Details
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019824.3 | NP_062798.1 | 4 | 259 | ||
GenBank mRNA | AB038243.1 | - | 3 | 195 | ||
AK003485.1 | - | 4 | 290 | |||
AK005437.1 | - | 4 | 278 | |||
AK171651.1 | - | 4 | 274 | |||
BC013618.1 | - | 4 | 259 | 4013537 | ||
BC054440.1 | - | 4 | 226 | 6395280 | ||
BC055036.1 | - | 4 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
