siRNA Details
Gene Symbol: Kcnj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168354.1 | NP_001161826.1 | 2 | 837 | ||
NM_019659.3 | NP_062633.1 | 2 | 835 | |||
GenBank mRNA | AF012834.1 | - | 2 | 656 | ||
BC020525.1 | - | 2 | 837 | 4241262 |
