siRNA Details
Gene Symbol: Sssca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020491.5 | NP_065237.3 | 3 | 276 | ||
GenBank mRNA | AB012865.1 | - | 3 | 244 | ||
AK005265.1 | - | 3 | 248 | |||
AK011957.1 | - | 3 | 266 | |||
AK012592.1 | - | 3 | 254 | |||
AK012704.1 | - | 3 | 254 | |||
BC021593.1 | - | 3 | 246 | 5102521 | ||
BY081504.1 | - | 3 | 334 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109537.
Gene Ontology Categories:
