siRNA Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019825.3 | NP_062799.2 | 10 | 4666 | ||
GenBank mRNA | AF216186.1 | - | 9 | 4551 | ||
AK129079.1 | - | 3 | 1825 | |||
AK142566.1 | - | 4 | 2851 | |||
AK171864.1 | - | 10 | 4824 | |||
BC083187.1 | - | 10 | 4666 | 30544739 | ||
JF707635.1 | - | 9 | 4525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
