siRNA Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242558.1 | NP_001229487.1 | 8 | 2893 | ||
NM_019825.3 | NP_062799.2 | 8 | 2893 | |||
GenBank mRNA | AF135169.1 | - | 1 | 391 | ||
AF216186.1 | - | 7 | 2778 | |||
AK129079.1 | - | 1 | 52 | |||
AK142566.1 | - | 2 | 1078 | |||
AK171864.1 | - | 8 | 3051 | |||
BC083187.1 | - | 8 | 2893 | 30544739 | ||
JF707634.1 | - | 7 | 2752 | |||
JF707635.1 | - | 7 | 2752 | |||
JF707636.1 | - | 7 | 2752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
