siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038587.4 | NP_001033676.2 | 2 | 1267 | ||
NM_001146296.1 | NP_001139768.1 | 2 | 1259 | |||
NM_019655.3 | NP_062629.3 | 2 | 1259 | |||
GenBank mRNA | AF052506.1 | - | 2 | 1172 | ||
AF291050.1 | - | 2 | 1166 | |||
AF291876.1 | - | 2 | 1166 | |||
AK076413.1 | - | 2 | 1218 | |||
AK089451.1 | - | 2 | 1230 | |||
AK147614.1 | - | 2 | 1230 | |||
BC042505.1 | - | 2 | 1193 | 4012994 | ||
CT010323.1 | - | 2 | 1166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
