siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038587.4 | NP_001033676.2 | 2 | 1347 | ||
NM_001146296.1 | NP_001139768.1 | 2 | 1339 | |||
NM_019655.3 | NP_062629.3 | 2 | 1339 | |||
GenBank mRNA | AF052506.1 | - | 2 | 1252 | ||
AK076413.1 | - | 2 | 1298 | |||
AK089451.1 | - | 2 | 1310 | |||
AK147614.1 | - | 2 | 1310 | |||
BC042505.1 | - | 2 | 1273 | 4012994 | ||
CT010323.1 | - | 2 | 1246 |

More Information
Other Species Targeted

Gene Ontology Categories:
