siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038587.4 | NP_001033676.2 | 13 | 3242 | ||
NM_001146296.1 | NP_001139768.1 | 13 | 3234 | |||
NM_019655.3 | NP_062629.3 | 13 | 3156 | |||
GenBank mRNA | AF052506.1 | - | 13 | 3069 | ||
AF291050.1 | - | 13 | 3063 | |||
AF291875.1 | - | 12 | 1631 | |||
AF291876.1 | - | 13 | 3141 | |||
AF291877.1 | - | 12 | 1709 | |||
AK076413.1 | - | 13 | 3193 | |||
AK089451.1 | - | 13 | 3205 | |||
AK147614.1 | - | 13 | 3205 | |||
AK156093.1 | - | 12 | 1723 | |||
AK172523.1 | - | 12 | 1723 | |||
BC042505.1 | - | 13 | 3090 | 4012994 | ||
CT010323.1 | - | 13 | 3063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
