siRNA Details
Gene Symbol: Ykt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019661.4 | NP_062635.2 | 5 | 584 | ||
GenBank mRNA | AF076956.1 | - | 5 | 456 | ||
AK002916.1 | - | 5 | 562 | |||
AK007486.1 | - | 5 | 552 | |||
AK147098.1 | - | 5 | 544 | |||
AK150472.1 | - | 5 | 541 | |||
AK159253.1 | - | 5 | 531 | |||
AK211548.1 | - | 2 | 98 | |||
BC006760.1 | - | 5 | 510 | 3257673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
