siRNA Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019674.3 | NP_062648.1 | 5, 6 | 425 | ||
GenBank mRNA | AF088911.1 | - | 5, 6 | 450 | ||
AK027923.1 | - | 3, 4 | 204 | |||
AK144244.1 | - | 4, 5 | 352 | |||
BC001993.1 | - | 5, 6 | 425 | 3491568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
