siRNA Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019674.3 | NP_062648.1 | 5, 6 | 416 | ||
GenBank mRNA | AF088911.1 | - | 5, 6 | 441 | ||
AK027923.1 | - | 3, 4 | 195 | |||
AK144244.1 | - | 4, 5 | 343 | |||
BC001993.1 | - | 5, 6 | 416 | 3491568 |
