siRNA Details
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 7 | 822 | ||
GenBank mRNA | AB028853.1 | - | 7 | 823 | ||
AK031858.1 | - | 7 | 889 | |||
AK077672.1 | - | 7 | 893 | |||
AK083208.1 | - | 7 | 890 | |||
AK163255.1 | - | 7 | 821 | |||
AK169169.1 | - | 7 | 882 | |||
AK211376.1 | - | 1 | 44 | |||
BC003420.1 | - | 7 | 895 | 2582183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
