siRNA Details
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011733.2 | NP_035863.1 | 2, 3 | 493 | ||
NM_139117.2 | NP_620817.2 | 2, 3 | 493 | |||
GenBank mRNA | AF246224.1 | - | 2, 3 | 335 | ||
AF248546.1 | - | 2, 3 | 315 | |||
AF248547.1 | - | 2, 3 | 315 | |||
AK003145.1 | - | 2, 3 | 427 | |||
AK076041.1 | - | 2, 3 | 432 | |||
BC027785.1 | - | 2, 3 | 504 | 5345031 | ||
BC048242.1 | - | 2, 3 | 481 | 6493766 | ||
BC062377.1 | - | 2, 3 | 481 | 6530404 | ||
D14485.1 | - | 2, 3 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
