siRNA Details
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019698.2 | NP_062672.2 | 15, 16 | 2052 | ||
NM_153554.2 | NP_705782.2 | 15, 16 | 2046 | |||
GenBank mRNA | AF056573.1 | - | 15, 16 | 1957 | ||
AF056574.1 | - | 15, 16 | 1951 | |||
AK134148.1 | - | 15, 16 | 2012 | |||
AK147711.1 | - | 15, 16 | 1972 | |||
AK151072.1 | - | 15, 16 | 1974 | |||
AK153946.1 | - | 15, 16 | 1973 | |||
AK159610.1 | - | 15, 16 | 1974 | |||
AK166459.1 | - | 15, 16 | 1938 | |||
AK166494.1 | - | 15, 16 | 1939 | |||
AK168905.1 | - | 15, 16 | 1972 | |||
AK190834.1 | - | 1, 2 | 44 | |||
BC033427.1 | - | 15, 16 | 1983 | 5009115 | ||
BC037699.1 | - | 15, 16 | 1931 | 4188900 |
