siRNA Details
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019698.2 | NP_062672.2 | 16 | 2065 | ||
NM_153554.2 | NP_705782.2 | 16 | 2059 | |||
GenBank mRNA | AF056573.1 | - | 16 | 1970 | ||
AF056574.1 | - | 16 | 1964 | |||
AK134148.1 | - | 16 | 2025 | |||
AK147711.1 | - | 16 | 1985 | |||
AK151072.1 | - | 16 | 1987 | |||
AK153946.1 | - | 16 | 1986 | |||
AK159610.1 | - | 16 | 1987 | |||
AK168905.1 | - | 16 | 1985 | |||
AK190834.1 | - | 2 | 57 | |||
BC033427.1 | - | 16 | 1996 | 5009115 | ||
BC037699.1 | - | 16 | 1944 | 4188900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
