siRNA Details
Gene Symbol: Kcnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111331.1 | NP_001104801.1 | 5 | 841 | ||
NM_001291005.1 | NP_001277934.1 | 6 | 663 | |||
NM_019789.4 | NP_062763.2 | 6 | 667 | |||
GenBank mRNA | AF184624.1 | - | 6 | 615 | ||
AF274050.1 | - | 5 | 527 | |||
AF287732.1 | - | 6 | 641 | |||
AF287733.1 | - | 6 | 645 | |||
AF300870.1 | - | 6 | 590 | |||
AK016721.1 | - | 6 | 829 | |||
AK165711.1 | - | 6 | 615 | |||
BC026980.1 | - | 6 | 604 | 5368268 | ||
BC047139.1 | - | 6 | 604 | 6389719 | ||
BC057329.1 | - | 6 | 586 | 6813819 | ||
DQ148499.1 | - | 5 | 527 | |||
DQ148500.1 | - | 5 | 841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
