siRNA Details
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019861.1 | NP_063914.1 | 3, 4 | 542 | ||
GenBank mRNA | AF136280.1 | - | 3, 4 | 443 | ||
AF197480.1 | - | 3, 4 | 487 | |||
AJ131851.1 | - | 3, 4 | 419 | |||
AK017474.1 | - | 3, 4 | 428 | |||
AK075862.1 | - | 3, 4 | 435 | |||
BC004054.1 | - | 13 | ||||
BC051131.1 | - | 2, 3 | 661 | |||
BC058758.1 | - | 3, 4 | 433 | 6492834 |
