siRNA Details
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019663.3 | NP_062637.2 | 7 | 925 | ||
GenBank mRNA | AF077950.1 | - | 7 | 917 | ||
AK013753.1 | - | 3 | 260 | |||
AK035682.1 | - | 1 | 354 | |||
AK075708.1 | - | 7 | 926 | |||
AK153871.1 | - | 7 | 930 | |||
BC051417.1 | - | 7 | 922 | 6515027 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106829, AY644724.
Gene Ontology Categories:
