siRNA Details
Gene Symbol: Nxt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110159.1 | NP_001103629.1 | 3 | 527 | ||
NM_019761.6 | NP_062735.4 | 2 | 418 | |||
GenBank mRNA | AF156958.2 | - | 2 | 359 | ||
AK003254.1 | - | 2 | 394 | |||
AK168314.1 | - | 2 | 378 | |||
BC023672.1 | - | 2 | 376 | 5039139 | ||
CJ186513.1 | - | 2 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
