siRNA Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019806.5 | NP_062780.2 | 4 | 455 | ||
GenBank mRNA | AF115504.1 | - | 4 | 333 | ||
AK050632.1 | - | 4 | 404 | |||
AK081908.1 | - | 4 | 384 | |||
AK160925.1 | - | 4 | 298 | |||
AK172080.1 | - | 4 | 297 | |||
BC048231.1 | - | 4 | 455 | 6313869 | ||
BC057123.1 | - | 4 | 444 | 6411688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
