siRNA Details
Gene Symbol: Cldn18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194921.1 | NP_001181850.1 | 5 | 806 | ||
NM_001194922.1 | NP_001181851.1 | 5 | 881 | |||
NM_001194923.1 | NP_001181852.1 | 5 | 818 | |||
NM_019815.3 | NP_062789.1 | 5 | 869 | |||
GenBank mRNA | AA028634.1 | - | 3 | 321 | ||
AF221068.1 | - | 5 | 821 | |||
AF349450.1 | - | 5 | 827 | |||
AF349451.1 | - | 5 | 807 | |||
AF349453.1 | - | 5 | 819 | |||
AK033657.1 | - | 5 | 807 |

More Information
Other Species Targeted

Gene Ontology Categories:
