siRNA Details
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019656.3 | NP_062630.2 | 4, 5 | 585 | ||
GenBank mRNA | AF053454.1 | - | 4, 5 | 532 | ||
AK146673.1 | - | 4, 5 | 490 | |||
AK149204.1 | - | 4, 5 | 495 | |||
AK160313.1 | - | 4, 5 | 479 | |||
AK167384.1 | - | 4, 5 | 481 | |||
BC003733.1 | - | 4, 5 | 519 | 3590862 | ||
BC110314.1 | - | 4, 5 | 494 | 5344878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
