siRNA Details
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019986.3 | NP_064370.2 | 6 | 998 | ||
GenBank mRNA | AF227684.1 | - | 6 | 941 | ||
AK016800.1 | - | 6 | 1041 | |||
AK144206.1 | - | 6 | 982 | |||
AK146525.1 | - | 6 | 971 | |||
BC082806.1 | - | 6 | 1000 | |||
BC119460.2 | - | 5 | 494 | |||
BC148304.2 | - | 5 | 502 |

More Information
Other Species Targeted

Gene Ontology Categories:
