siRNA Details
Gene Symbol: Sec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF113532.1 | - | 1 | 458 | ||
BC103557.1 | - | 1 | 457 | |||
BC103558.1 | - | 1 | 457 | |||
BC103559.1 | - | 1 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
