siRNA Details
Gene Symbol: Txn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019913.5 | NP_064297.1 | 2 | 295 | ||
GenBank mRNA | AK002358.1 | - | 2 | 270 | ||
AK010917.1 | - | 2 | 296 | |||
AK090199.1 | - | 2 | 276 | |||
AK147164.1 | - | 1 | 1223 | |||
AK149855.1 | - | 2 | 279 | |||
AK160591.1 | - | 2 | 298 | |||
AK167754.1 | - | 2 | 261 | |||
AK167925.1 | - | 2 | 233 | |||
AK168322.1 | - | 2 | 261 | |||
BC068182.1 | - | 2 | 281 | 30633447 | ||
U85089.1 | - | 2 | 258 |

More Information
Other Species Targeted

Gene Ontology Categories:
