siRNA Details
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027044.3 | NP_081320.2 | 2, 3 | 245 | ||
GenBank mRNA | AF108357.1 | - | 2, 3 | 205 | ||
AK004438.1 | - | 2, 3 | 200 | |||
AK005682.1 | - | 2, 3 | 191 | |||
AK028206.1 | - | 2, 3 | 182 | |||
AK145883.1 | - | 2, 3 | 198 | |||
AK211903.1 | - | 1, 2 | 44 | |||
BC026920.1 | - | 2, 3 | 182 | 3666296 | ||
CN835522.1 | - | 2, 3 | 248 |

More Information
Other Species Targeted

Gene Ontology Categories:
