siRNA Details
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019924.2 | NP_064308.2 | 4 | 447 | ||
GenBank mRNA | AF074714.1 | - | 4 | 419 | ||
AK089467.1 | - | 4 | 438 | |||
AK154674.1 | - | 4 | 446 | |||
BC012964.1 | - | 4 | 457 | 4160873 | ||
CJ065157.1 | - | 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
