siRNA Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019946.4 | NP_064330.2 | 2 | 152 | ||
GenBank mRNA | AF159050.1 | - | 1 | 51 | ||
AK002800.1 | - | 2 | 153 | |||
AK005122.1 | - | 2 | 153 | |||
BC009155.1 | - | 1 | 91 | 2655738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
