siRNA Details
Gene Symbol: Crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019922.2 | NP_064306.2 | 3, 4 | 901 | ||
GenBank mRNA | AJ006469.1 | - | 3, 4 | 859 | ||
AK007624.1 | - | 3, 4 | 622 | |||
AK017797.1 | - | 3, 4 | 864 | |||
AK047506.1 | - | 3, 4 | 857 | |||
BC049890.1 | - | 3, 4 | 838 | 6306169 |

More Information
Other Species Targeted

Gene Ontology Categories:
