siRNA Details
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162989.1 | NP_001156461.1 | 2 | 720 | ||
NM_019996.4 | NP_064380.3 | 2 | 830 | |||
GenBank mRNA | AJ276504.1 | - | 2 | 661 | ||
AK016958.1 | - | 2 | 722 | |||
AK030749.1 | - | 2 | 688 | |||
BC016530.1 | - | 2 | 677 | |||
BC049590.1 | - | 2 | 676 | 6587693 | ||
BC061110.1 | - | 2 | 690 | 30303763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
